missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate Heptahydrate, ACS Reagent Grade, Scharlab
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
- Synonyms: Bitter salt, Epsom salt, Sulfuric acid magnesium salt heptahydrate
- Molecular Formula: MgSO4 • 7H2O
- Molar Mass: 246.48g/mol
- CAS: 10034-99-8
- EIN ECS No.: 231-298-2
- Loss on Ignition: 48–52% (400°C)
Specifications
Specifications
CAS | 10034-99-8 |
pH | 5.0–8.2 (5%, water) |
Quantity | 500 g |
Molecular Formula | H14MgO11S |
MDL Number | MFCD00149785 |
Synonym | magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate |
InChI Key | WRUGWIBCXHJTDG-UHFFFAOYSA-L |
SMILES | O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O |
IUPAC Name | magnesium(2+) heptahydrate sulfate |
Molecular Weight (g/mol) | 246.47 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Magnesium Sulfate Heptahydrate, ACS Reagent Grade, Scharlab >
Spot an opportunity for improvement?Share a Content Correction