missing translation for 'onlineSavingsMsg'
Learn More
Magnesium sulfate heptahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals Product Code.: 11357337

Magnesium sulfate heptahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Product Code. 11357337
2 kg
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11357337

Brand: Thermo Scientific Alfa Aesar 011596.A3

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Fireproofing, Textiles, Catalyst Carrier

Magnesium sulfate produced by appropriate process finds use in several pharmaceutical preparations of magnesium, usually known as epsom salt. It is used as a micronutrient in agriculture, brewing salt in production of certain alcoholic products, electrolyte to prepare metal salts, coagulant, and ingredient in bath salts for use in floatation therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Crystals
Quantity 2 kg
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in in water and acid. Slightly soluble in alcohol and glycerol.
Formula Weight 246.48 (120.37 Anhydrous)
Grade ACS
Odor Odorless
Appearance White crystals
Density 1.68
Chemical Name or Material Magnesium sulfate heptahydrate, heptahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN13L009 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog Number011596.22,011596.36,011596.A3
Lot NumberM28L038 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number011596.22,011596.36,011596.A3
Lot NumberU19K030 Certificate TypeCertificate of Analysis Date20/09/2024 Catalog Number011596.22,011596.36,011596.A3
Lot NumberQ09K043 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog Number011596.22,011596.36,011596.A3
Lot NumberT05J037 Certificate TypeCertificate of Analysis Date13/07/2023 Catalog Number011596,011596.22,011596.36,011596.A3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Magnesium sulfate heptahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.