Learn More
Magnesium rod, 7.9mm (0.31in) dia x 25mm (1in) length, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 15486535
Magnesium rod, 7.9mm (0.31in) dia x 25mm (1in) length, 99.9% (metals basis), Thermo Scientific Chemicals
Product Code. 15486535
5 Pc., Pack of 5
Click to view available options
Quantity:
1 Pc.
25 Pc.
5 Pc.
Unit Size:
Each
Pack of 25
Pack of 5
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15486535
Brand: Thermo Scientific Alfa Aesar 043355.KJ
Request Bulk or Custom Format
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]
Specifications
Quantity 5 Pc.
Percent Purity 99.9%
Odor Odorless
Weight ≈2.1g/piece
Form Rod
Assay (metals basis)
Chemical Name or Material Magnesium rod, 7.9mm (0.31 in.) dia. x 25mm (1 in.) length
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberM21H055 Certificate TypeCertificate of Analysis Date17/10/2024 Product Code043355.KJ,043355.KM,043355.KF
Lot NumberW29D011 Certificate TypeCertificate of Analysis Date04/04/2024 Product Code043355.KJ,043355.KM,043355.KF
Lot NumberF11T027 Certificate TypeCertificate of Analysis Date07/06/2023 Product Code043355.KJ
Lot NumberR03I026 Certificate TypeCertificate of Analysis Date24/05/2022 Product Code043355.KJ,043357.22,043357.06,043355.KM,043355.KF
Lot NumberL10L39 Certificate TypeCertificate of Analysis Date12/11/2021 Product Code043355.KJ,043357.22,043357.06,043355.KM,043355.KF
Product Title
Magnesium rod, 7.9mm (0.31in) dia x 25mm (1in) length, 99.9% (metals basis), Thermo Scientific Chemicals >