Learn More
Applied Biosystems™ MagMAX™ Viral/Pathogen Binding Solution
This binding buffer is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.
Brand: Applied Biosystems™ A42359
Description
This binding buffer is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42352, A42356, A42357, and A42358). It is made available separately for applications that require more binding buffer than is provided in the kit.
Specifications
Store at room temperature. |
|
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR | |
MagMAX™ | |
550 mL | |
Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids |
Bacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Virus, Yeast | |
High-throughput Compatible, Manual Protocols, Automated Protocols | |
Viral/Pathogen Binding Solution | |
Room Temperature |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
For Research Use Only. Not for use in diagnostic procedures.