missing translation for 'onlineSavingsMsg'
Learn More
m-Toluic acid, 99% Product Code.: 10573111

m-Toluic acid, 99%

Product Code. p-200994 Shop All Thermo Scientific Chemicals Products
5 g, Glass Bottle, 5g
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
1kg
5g
This item is not returnable. View return policy

Product Code. 10573111

Brand: Thermo Scientific Acros 139050050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-04-7 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-04-7
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002523
InChI Key GPSDUZXPYCFOSQ-UHFFFAOYSA-N
Synonym m-toluic acid, m-toluylic acid, 3-toluic acid, benzoic acid, 3-methyl, meta-toluic acid, m-methylbenzoic acid, beta-methylbenzoic acid, 3-methyl-benzoic acid, beta-bethylbenzoic acid, unii-1ua7k8eext
PubChem CID 7418
ChEBI CHEBI:10589
IUPAC Name 3-methylbenzoic acid
SMILES CC1=CC=CC(=C1)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 108°C to 112°C
Color Beige-Yellow to Yellow
Density 1.05
Boiling Point 263°C
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula CH3C6H4CO2H
Quantity 5 g
Beilstein 09, 475
Merck Index 14, 9535
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 136.15
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material m-Toluic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
m-Toluic acid, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.