Documents
Product Information
Manuals
- 1
- 2
Spin up to six microplates and, using sealed carriers, ensure certified sealed conditions for all samples with the Thermo Scientific™ M-20 Microplate Swinging-Bucket Rotor.
Brand: Thermo Scientific™ 75003624
21113.02 NOK valid until 2025-12-31
Use promo code "24378" to get your promotional price.
Spin up to six microplates and, using sealed carriers, ensure certified sealed conditions for all samples with the Thermo Scientific™ M-20 Microplate Swinging-Bucket Rotor.
Ordering Information:
Order 75003625 for Sealed Carriers, with trays and lids (set of 2), and 75002011 for Replacement Sealing Lids (set of 2). For Replacement O-rings for lids (set of 4 with grease) order 75002012. Contact your Sales Representative for information.
Includes:
Two buckets, two adapters
Compatible with:
Thermo Scientific Sorvall™ 16/16R, Sorvall Lengend™ X1/X1R, Multifuge™ X1, Megafuge™ 16 Centrifuge Series
M-20 Microplate Rotor | |
2 x 3 Standard or 2 x 1 Deepwell Plates | |
4,000 rpm | |
Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Sorvall ST 40, Sorvall ST 16, Multifuge X3/X3F, Multifuge X1, Megafuge 40, Megafuge 16 Centrifuge Series | |
Biocontainment certified by CAMR™, HPA, Porton Down, U.K. |
Swinging Bucket Rotor | |
2,272 x g | |
12.7 cm | |
90° | |
Yes |
Expires: 2025-12-31
Expires: 2025-06-30
Expires: 2025-08-24
Expires: 2025-07-31