missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24899-1-AP-150UL
This item is not returnable.
View return policy
Description
LDL related proteins, LRP 5 and LRP 6, are members of the LDL receptor superfamily, which were found to be involved in the canonical Wnt signaling pathway. Mammalian LRP 6 was shown to bind to Wnt 1 and enhance Wnt-induced developmental processes in Xenopus embryos. Mice lacking LRP 6 exhibited developmental defects that are similar to those caused by deficiencies in various Wnt proteins. Recent work also shows that Wnt proteins induce the binding of LRP 5 to Axin, leading Axin degradation and beta catenin stabilization.Specifications
LRP5 | |
Polyclonal | |
Unconjugated | |
LRP5 | |
BMND1, EVR1, EVR4, HBM, LR3, LRP 5, LRP5, LRP7, OPPG, OPS, OPTA1, VBCH2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4041 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75197 | |
LRP5 | |
LRP5 Fusion Protein Ag19034 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction