Learn More
IKA™ LR 2000.1 Reactor Vessel
Use this vessel for the optimization and reproduction of various chemical reactions, mixing and homogenization processes. IKA™ LR 2000.1 Reactor Vessel features double walled glass and quick-action connectors.
Brand:  IKA™ 0002508300
49200.98 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 10183041
  • 51790.50 NOK / Each
Price-Shattering Deals on Lab Glassware
Use promo code 24123 at checkout
View Available Promotions
Only null left
Estimated Shipment: 20-08-2025
Description
For use with IKA LR-2.ST and LR 2000 V systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For LR-2.ST and LR 2000 V
Reactor Vessel
Certificates
Promotions
Promotions
Product Content Correction
Product Title
IKA™ LR 2000.1 Reactor Vessel > Reactor Vessel; For Use With LR-2.ST and LR 2000 V