missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loxiglumide
CCK1 Receptor Antagonists
505.39 NOK - 2118.74 NOK
Specifications
CAS | 107097-80-3 |
---|---|
Synonym | 4-[(3,4-Dichlorobenzoyl)amino]-5-[(3-methoxypropyl)pentylamino]-5-oxopentanoic acid |
Molecular Formula | C21H30Cl2N2O5 |
Formula Weight | 461.38 |
Purity | 97% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780809
|
Tocris Bioscience
3036/5 |
5 mg |
505.39 NOK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18767108
|
Tocris Bioscience
3036/25 |
25 mg |
2118.74 NOK
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCK1 antagonist. Suppresses CCK-induced proliferation of acinar cells. Inhibits pancreatic secretion of digestive enzymes. Active in vivo.Specifications
107097-80-3 | |
C21H30Cl2N2O5 | |
97% |
4-[(3,4-Dichlorobenzoyl)amino]-5-[(3-methoxypropyl)pentylamino]-5-oxopentanoic acid | |
461.38 |
Safety and Handling
Recommended Storage : Store at -20°C