Learn More
Cytiva Lower Section
This Lower Section is a replacement part for Whatman™ GV 050 Series Vacuum Filtration Manifolds.
Brand: Cytiva 10442002
756.87 NOK valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Description: Replacement Part for GV 050 Series Glass Vacuum Filtration Manifolds
- For Use With (Application): Vacuum Filtration
Specifications
Autoclavable | |
Reusable | |
Borosilicate Glass | |
Vacuum Filtration | |
1 Ea. |
Filter Funnel (Base Section) | |
Whatman GV050/0 Glass Vacuum Filter Holder | |
Non-sterile | |
GV050/0/02 | |
Filter Funnel |