missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Low Iodine Test Diet, Powder

Low in iodine diet.

Brand:  MP Biomedicals 0290462250

 View more versions of this product

Product Code. 11465752

  • 20077.46 NOK / 50kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Low in iodine diet
  • Contains yellow corn and menadione
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Single-use
50 kg
Room Temperature
0.3048 m
Animal Diet
Powder
Non-sterile
1 ft.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.