missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Low Iodine Diet, Fortified With Vitamins, Pellet

Vitamin fortified diet, low in iodine.

8059.18 NOK - 34993.78 NOK

Specifications

Product Type Animal Diet
Autoclavable Autoclavable
Disposable Single-use
Format Pellet
Sterility Non-sterile
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11435592
View Documents
MP Biomedicals
0290220410
10 kg
8059.18 NOK
10kg
Please to purchase this item. Need a web account? Register with us today!
11445592
View Documents
MP Biomedicals
0290220420
20 kg
16096.10 NOK
20kg
Please to purchase this item. Need a web account? Register with us today!
11455592
View Documents
MP Biomedicals
0290220450
50 kg
34993.78 NOK
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • This diet is low in iodine but fortified with other vitamins
  • Ideal for research on iodine deficiency diets
Specifications

Specifications

Animal Diet
Single-use
Non-sterile
1 ft.
Autoclavable
Pellet
Room Temperature
0.3048 m
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.