Learn More
Kartell™ Low Form Deep Tray with Draining Holes
Rectangular tank with draining holes on all sides. Stackable with recessed rim to allow ventilation.
Brand:  Kartell™ 0054700
Product Code. 15666395
  • 464.00 NOK / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Description
  • Draining holes on all sides
  • Handles
  • Stackable
  • Recessed rim for ventilation
  • High-density polyethylene (HDPE)
  • Specifications
    Specifications
    16 L
    White
    350 mm
    Deep Tray
    Rectangular
    350 mm x 540 mm
    115 mm
    Polyethylene
    540 mm