missing translation for 'onlineSavingsMsg'
Learn More
Lotamilast, MedChemExpress Product Code.: 16401517

Lotamilast, MedChemExpress

Product Code. 16401517
50 mg, 50mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16401517

Brand: Medchem Express HY12740

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lotamilast (RVT-501; E6005) is a selective phosphodiesterase 4 (PDE4) inhibitor with an IC50 of 2.8 nM.

Lotamilast (RVT-501

Specifications

Chemical Name or Material Lotamilast
CAS 947620-48-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H24N4O5
Quantity 50 mg
Synonym RVT-501 E6005
Solubility Information DMSO : 25 mg/mL (52.91 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C(OC)=O)C=C1)NC2=CC=CC(C3=C4C(C=C(OC)C(OC)=C4)=NC(NC)=N3)=C2
Molecular Weight (g/mol) 472.49
Formula Weight 472.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Lotamilast, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.