Learn More
SeaKem™ HGT Agarose, Lonza™
  • Used in counterimmunoelectrophoresis and crossed immunoelectrophoresis
  • High gelling temperature (42°C ± 1.5°C)
  • High gel strength (≥1000g/cm2) and clarity
2700.00 NOK - 9105.00 NOK
Specifications
Chemical Name or Material SeaKem HGT Agarose
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11597775
View Documents Promotion Details
Lonza
50041
25 g
2700.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 09-10-2024
to see stock.
Add to basket
10407885
View Documents Promotion Details
Lonza
50040
125 g
9105.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-10-2024
Specifications
SeaKem HGT Agarose