Learn More
Lonza™ Reliant™ Precast RNA Gels
Formulated for separation of RNA of sizes from 0.25 to at least 10kb. Reliant™ Precast RNA Gels are ready-to-use RNase free agarose gels suitable for a variety of RNA applications including Northern blotting and analysis of RNA transcripts.
5000.00 NOK
Specifications
Concentration 1.25%
Format Multiwell
Product Type Precast Gel
View More Specs
Products 2
Product Code Brand Format Price Quantity & Availability  
Product Code Brand Format Price Quantity & Availability  
11537825
View Documents Promotion Details
Lonza™
54922
Multiwell
5000.00 NOK
Pack of 20
Estimated Shipment: 14-11-2024
to see stock.
Add to basket
11577835
View Documents Promotion Details
Lonza™
54948
Multiwell
5000.00 NOK
Each
Estimated Shipment: 14-11-2024
Description
  • Ready to use
  • RNase free
  • Free of denaturants, eliminating problems associated with staining gels containing formaldehyde
  • Suitable for a variety of applications, including Northern blotting and analysis of RNA transcripts
  • Compatible with most common mini chambers
  • Available with a separate container of glyoxal buffer
Specifications
Specifications
1.25%
Multiwell
Precast Gel