Learn More
Lonza™ BioWhittaker™ X-VIVO™ 10 Hematopoietic Serum-Free Culture Media
For adoptive immunotherapy and cellular therapy research
Brand: Lonza™ BE04-743Q
This item is not returnable.
View return policy
Description
- Does not contain any lymphokines required for activation of human lymphocytes
- Intended for in vitro procedures
- Device Master Records (DMRs) for these products are held in an FDA Master File
- Chemically defined media optimized for specific cell types
- For further manufacturing or laboratory use
- Designed to support the generation of lymphokine activated killer (LAK) cells in a serum-free environment
- Limited by Federal (USA) law to investigational use
- These products are for research use only
- Not approved for human or veterinary use, for application to humans or animals, or for use in clinical or in vitro procedures
Specifications
Stem Cell Serum Free Media (SFM) | |
No Gentamicin, No Phenol Red | |
1 L |
Product Content Correction
Product Title
Lonza™ BioWhittaker™ X-VIVO™ 10 Hematopoietic Serum-Free Culture Media > X-VIVO 10; w/o Gentamicin and Phenol Red, 1L