missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ Versene™ (EDTA), 0.02%
It is a chelating agent used to lift some cell types from the culture vessel surface into suspension
Brand: Lonza™ BE17-711E
This item is not returnable.
View return policy
Description
- Manufactured in accordance with cGMP regulations
- Cell culture tested to assure compatibility
Specifications
15° to 30°C | |
No Calcium, No Magnesium | |
100 mL |
Cell Culture Dissociation Reagent | |
7.08 to 7.5 |