Learn More
Lonza™ BioWhittaker™ ProNSO™ CD Media
Chemically defined, protein-free medium designed to meet a growing demand for optimized NS0 formulations
Description
These NS0 products provides maximum protein productivity. Two distinct proprietary media formulations are offered to cover a broad range of nutritional needs; try each to determine which one provides the best yield for your application.
- Further maximize protein production by titration with ProNS0 Lipid CD Supplement
- Formulations contain HEPES and Pluronic, without L-glutamine, phenol red, or cholesterol
- Product purification is simplified
- Lot-to-lot consistency ensures dependable performance
- Non-animal origin components reduce regulatory burdens
Specifications
Liquid | |
ProNS0™ |
Product Title
Lonza™ BioWhittaker™ ProNSO™ CD Media