Learn More
Lonza™ BioWhittaker™ ProCHO Media, Media
Brand: Lonza™ BE02-016Q
This item is not returnable.
View return policy
Description
For adherent culture of CHO cells. Contains L-glutamine. Does not contain hypoxanthine or thymidine.
Specifications
2° to 8°C | |
Liquid | |
1 L |
CHO | |
No Hypoxanthine, No Thymidine |