missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ Insect Cell Culture Media: Grace's Insect Medium
Chemically-defined media optimized for specific cell types
Brand: Lonza™ 04-457F
This item is not returnable.
View return policy
Description
Compliance
Manufactured in accordance with cGMP regulations for insect cell lines; formulations recommended by the Tissue Culture Association.
Specifications
2° to 8°C | |
Grace's Insect Medium | |
No Lactalbumin Hydrolysate, No Yeastolate, No Gentamicin |
Liquid | |
None | |
500 mL |