Learn More
Lonza™ BioWhittaker™ Hybridoma Serum-Free and Protein-Free Culture Media: HL-1 Supplement (100X)
Prescreened in actual hybridoma systems to ensure minimal variation
Brand: Lonza™ 77227
This item is not returnable.
View return policy
Description
- HL-1 supports the serum-free growth of hybridomas and differentiated cells of lymphoid origin
1Rehydrates to quantity
Specifications
2° to 8°C | |
Liquid | |
10 mL |