Learn More
Lonza™ BioWhittaker™ Hybridoma Serum-Free and Protein-Free Culture Media: HL-1 Supplement (100X)
Prescreened in actual hybridoma systems to ensure minimal variation
Brand:  Lonza™ 77227
Product Code. 11650121
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • HL-1 supports the serum-free growth of hybridomas and differentiated cells of lymphoid origin
1Rehydrates to quantity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2° to 8°C
Liquid
10 mL