missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Trypsin-Versene™-Trypsin-EDTA Cell Culture Reagent
Combined reagent used to remove cells from culture vessel surface into suspension.Trypsin-Versene™-Trypsin-EDTA Cell Culture Reagent aids in cell-cell adhesion.
Brand: Lonza™ BE17-161E
This item is not returnable.
View return policy
Description
- EDTA added to trypsin solutions to enhance activity
- Contains 0.5g/L Trypsin 1:250; 0.2g/L Versene (EDTA)
- Manufactured with irradiated trypsin
- Porcine parvovirus tested and mycoplasma screened
Specifications
-10° to -20°C | |
Sterile | |
100 mL |
Cell Culture Dissociation Reagent | |
7.1 to 7.9 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction