Learn More
Lonza™ BioWhittaker™ Antibiotics: Penicillin-Streptomycin Mixtures
Used as a broad spectrum bacteriostatic and bactericidal, with activity against gram negative and gram-positive organisms including yeast and mold. BioWhittaker™ Antibiotics: Penicillin-Streptomycin Mixtures are manufactured in accordance with cGMP regulations.
Brand: Lonza™ 17-745E
This item is not returnable.
View return policy
Description
- Suggested working concentration of Penicillin-Streptomycin is 100 units of Potassium Penicillin and 100 μg of Streptomycin Sulfate per 1 ml of culture media
- Manufactured in accordance with cGMP regulations
- Cell culture tested to assure compatibility.
- 20mL vials have crimp-top closure
- 4.5, 100 and 500mL bottles have screw-cap closure
Specifications
-10° to -20°C | |
10K/10K/25μg | |
100 mL |
Antibiotic-Antimycotic | |
3.3 to 7.2 |