missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ Antibiotics: Amphotericin B
Manufactured in accordance with cGMP regulations and cell culture tested to assure compatibility
Brand: Lonza™ 17-836R
This item is not returnable.
View return policy
Description
- 20mL vials have crimp-top closure
- 100mL bottles have screw-cap closure
Specifications
-10° to -20°C | |
250 μg/mL | |
Antifungal | |
20 mL |