Learn More
Hamilton™ Long Life Autosampler
Brand:  Hamilton™ 75830
Product Code. 10401973
  • 1335.00 NOK / Pack of 25
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Specifications
Yes
25 Pk.
3 mm
Product Title
Hamilton™ Long Life Autosampler > Diameter: 17mm