Learn More
Hirschmann™ Long Conical Centrifuge Tubes
Excellent for analysis and separation of various materials. Hirschmann Instruments™ Long Conical Centrifuge Tubes are manufactured to tolerate high speeds.
Brand:  Hirschmann™ 6000101
 View more versions of this product
Product Code. 13158019
  • 1400.00 NOK / Pack of 100
Only null left
Estimated Shipment: 16-07-2025
Description
  • Long conical form
  • Available with either white or no graduations
Specifications
Specifications
15 mL
115 ±5 mm
No
100
17 mm
Centrifuge Tube
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hirschmann™ Long Conical Centrifuge Tubes > Outer Diameter: 17mm