missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Tetraborate, Honeywell™
≥99.9% trace metals basis
1763.71 NOK - 5973.98 NOK
Chemical Identifiers
CAS | 12007-60-2 |
---|---|
Molecular Formula | B4Li2O7 |
Molecular Weight (g/mol) | 169.11 |
MDL Number | MFCD00011083 |
InChI Key | RDZCISUGRQUANA-UHFFFAOYSA-N |
IUPAC Name | dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) |
SMILES | [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15672190
|
Honeywell Research Chemicals
222534-250G |
250 g |
1763.71 NOK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15662190
|
Honeywell Research Chemicals
222534-1KG |
1 kg |
5973.98 NOK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
12007-60-2 | |
169.11 | |
RDZCISUGRQUANA-UHFFFAOYSA-N | |
[Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 |
B4Li2O7 | |
MFCD00011083 | |
dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) |
Specifications
12007-60-2 | |
Li2B4O7 | |
NONH for all modes of transport | |
[Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2 | |
169.11 | |
≥99.9% (trace metals basis) | |
Lithium Tetraborate |
B4Li2O7 | |
MFCD00011083 | |
RDZCISUGRQUANA-UHFFFAOYSA-N | |
dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate) | |
169.12g/mol | |
≥0.25g/mL at 25°C |
Safety and Handling
missing translation for 'einecsNumber' : 234-514-3
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lithium Tetraborate, Honeywell™