missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Sulfate Monohydrate, (ACS Reagent Grade), MP Biomedicals
Click to view available options
Quantity:
100 g
500 g
Specifications
Specifications
Quantity | 100 g |
Formula Weight | 128 |
Grade | ACS Reagent |
Chemical Name or Material | Lithium Sulfate Monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'