Learn More
Lithium methoxide, 98+% Product Code.: 11309618
Lithium methoxide, 98+%
Product Code. 11309618
25 g, 25g
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. 11309618
Brand: Thermo Scientific Alfa Aesar 044129.14
Request Bulk or Custom Format
CAS: 865-34-9 | CH3LiO | 37.97 g/mol
Lithium methoxide is used in sol-gel synthesis of lithium borophosphates. It is also useful for deprotonation reactions in organic synthesis. Further, it serves as a mild base in organic synthesis especially in transesterifications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 865-34-9
Molecular Formula CH3LiO
Molecular Weight (g/mol) 37.97
MDL Number MFCD00036357
InChI Key JILPJDVXYVTZDQ-UHFFFAOYSA-N
Synonym lithium methoxide, lithium methanolate, methanol, lithium salt, lithium methylate, liome, methanol, lithium salt 1:1, liom, lithium methylat, lithotab methoxide, lioch3Show MoreShow Less
PubChem CID 4177230
SMILES [Li+].C[O-]
Specifications
Melting Point >500°C (decomposition)
Color White
Linear Formula LiOCH3
Quantity 25 g
UN Number UN3206
Beilstein 3552389
Sensitivity Air and moisture sensitive
Solubility Information Soluble in methanol.
IUPAC Name lithium(1+) methanolate
Formula Weight 37.98
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Lithium methoxide
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberQ26F008 Certificate TypeCertificate of Analysis Date15/01/2025 Product Code044129.14,044129.36,044129.22
Lot NumberX09K051 Certificate TypeCertificate of Analysis Date05/11/2024 Product Code044129.14,044129.36,044129.22
Lot NumberU12K011 Certificate TypeCertificate of Analysis Date28/08/2024 Product Code044129.14,044129.36,044129.22
Lot NumberY17J028 Certificate TypeCertificate of Analysis Date05/01/2024 Product Code044129.14,044129.36,044129.22
Lot NumberB08L26 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code044129.14,044129.36,044129.22
Product Title
Lithium methoxide, 98+% >