missing translation for 'onlineSavingsMsg'
Learn More
Lithium Chloride, 99% (ACS Grade), MP Biomedicals Product Code.: 11402872

Lithium Chloride, 99% (ACS Grade), MP Biomedicals

Product Code. 11402872 Shop All MP Biomedicals Products
500 g
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy

Product Code. 11402872

Brand: MP Biomedicals 0215253890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7447-41-8
Quantity 500 g
Molecular Formula ClLi
MDL Number MFCD00011078
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES [Li+].[Cl-]
IUPAC Name lithium(1+) chloride
Molecular Weight (g/mol) 42.39
PubChem CID 433294
ChEBI CHEBI:48607
Formula Weight 42.4
Grade ACS
Chemical Name or Material Lithium Chloride, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium Chloride, 99% (ACS Grade), MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.