missing translation for 'onlineSavingsMsg'
Learn More

Lithium, 99+%, granular, dry

Product Code. p-222326 Shop All Thermo Scientific Chemicals Products
25 g, 25g
Click to view available options
Quantity:
25 g
250 g
Unit Size:
250g
25g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 7439-93-2 | Li | 6.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-93-2
Molecular Formula Li
Molecular Weight (g/mol) 6.94
MDL Number MFCD00134051
InChI Key WHXSMMKQMYFTQS-UHFFFAOYSA-N
Synonym litio, litium, lithium, metallic, lithium, elemental, monohydride, unii-9fn79x2m3f, hsdb 647, hydride lih, hydrure de french, hsdb 549
PubChem CID 3028194
ChEBI CHEBI:30145
IUPAC Name lithium
SMILES [Li]

Specifications

Melting Point 180.0°C
Boiling Point 1340.0°C
Density 0.5300g/mL
Assay Percent Range 99+%
Quantity 25 g
Fieser 01,570; 03,174; 04,286; 05,376; 10,234; 11,286; 13,157; 15,184; 17,161
Merck Index 15, 5576
Solubility Information Solubility in water: reacts. Other solubilities: forming lioh and hydrogen, soluble in liquid ammonia and acids, decomposes in alcohol
Formula Weight 6.94
Percent Purity ≥99%
Form Granular
Chemical Name or Material Lithium, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium, 99+%, granular, dry >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.