Learn More
Buerkle™ Liquid-Sampler, Place onto Bottle
The valve is opened by attaching the sample container to empty the sample. Sampler for thin to viscous liquids. Easy use with one-handed operation. With discharge funnel for safe filling of the sample bottles. Material AISI 316L (1.4404).
Brand:  Buerkle™ 5326-0003
 View more versions of this product
Product Code. 10397685
  • 3990.00 NOK / Each
Only null left
Estimated Shipment: 06-08-2025
Description
The valve is opened when the sampler is attached to the sample container. Comes complete with discharge funnel for safe filling of sample bottles.
  • V4A
  • Autoclavable up to max. 121°C 20 min.
Specifications
Specifications
Liquid Sampler
Autoclavable
Stainless Steel (V4A)
32 mm
Stainless Steel (V4A)
Discharge Funnel
50 mL
455 mm
Videos
  • Buerkle V4A Stainless Steel Liquid Sampler
    Buerkle V4A Stainless Steel Liquid Sampler
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Buerkle V4A Stainless Steel Liquid Sampler
    SDS
    Documents
    Documents
    Brochures
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    Buerkle™ Liquid-Sampler, Place onto Bottle > Capacity: 50mL