missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIPT2-AS1 Antibody (OTI4C7), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71949DL594
This item is not returnable.
View return policy
Description
LIPT2-AS1 Monoclonal antibody specifically detects LIPT2-AS1 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
LIPT2-AS1 | |
Monoclonal | |
DyLight 594 | |
LIPT2 antisense RNA 1, MGC12965 | |
Full length recombinant protein of human Cytochrome c (NP_940921) produced in E.coli. | |
0.1 mL | |
Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers | |
100287896 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI4C7 | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LIPT2-AS1 Antibody (OTI4C7), DyLight 594, Novus Biologicals™ > 0.1 mL; DyLight 594
Spot an opportunity for improvement?Share a Content Correction