Learn More
Gilac™ Lightweight Nesting Pallet Product Code.: 17988621
Estimated Shipment: 03-12-2025
  • ONLINE EXCLUSIVE PRICE
    457.80 NOK
    331.19 NOK / Each
    Save 126.61 NOK
    27% Off
Only null left
Add to basket
Gilac™ Lightweight Nesting Pallet
Product Code. p-200009589
600 x 800 x 130 mm, Each
Click to view available options
Dimensions (L x W x H):
600 x 800 x 130 mm
800 x 1200 x 155 mm
1200 x 1000 x 160 mm
Unit Size:
Each
Product Code. 17988621
Brand: Gilac™ G052204
  • ONLINE EXCLUSIVE PRICE
    457.80 NOK
    331.19 NOK / Each
    Save 126.61 NOK
    27% Off
Only null left
Estimated Shipment: 03-12-2025
Add to basket
Only null left
Add to basket
Lightweight Pallet is nesting and therefore space-saving when stored empty.
  • Lightweight nesting plastic pallet.
  • Nesting and therefore space-saving when stored empty.
  • Storage prohibited on racks: the pads of the pallets are incompatible with the rails of the racks.
Specifications
Product Type Aero Tote Pallet
Color Black
Dimensions (L x W x H) 600 x 800 x 130 mm
Capacity (English) 1543.24 lb.
Capacity (Metric) 700 kg
Height (English) 5.1 in.
Height (Metric) 130 mm
Length (English) 23.6 in.
Length (Metric) 600 mm
Width (English) 31.5 in.
Width (Metric) 800 mm
Product Title
Gilac™ Lightweight Nesting Pallet >