Learn More
Invitrogen™ Novex™ IL-10 Human Ultrasensitive Magnetic Bead Kit
Designed for quantification of low levels of human IL-10 in serum, plasma and tissue culture supernatant samples
Brand: Invitrogen™ LHC0103M
Includes: Antibody coated capture beads, standard, detector antibody and lot-specific technical data sheet