Learn More
Haldenwanger™ Lids for Porcelain Crucibles
Use these lids with porcelain crucibles. Haldenwanger™ Lids for Porcelain Crucibles are constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ 79D/9
 View more versions of this product
Product Code. 12346507
  • 657.20 NOK / Pack of 10
Only null left
Estimated Shipment: 04-08-2025
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Description
Description
  • Available in a variety of sizes
  • For use with Haldenwanger porcelain crucibles
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
Porcelain
28mm
Crucible Lid
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Haldenwanger™ Lids for Porcelain Crucibles > Diameter: 28mm