missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lidocaine Hydrochloride MP Biomedicals
Click to view available options
Quantity:
25 g
Specifications
Specifications
CAS | 6108-05-0 |
Molecular Formula | C14H25ClN2O2 |
MDL Number | MFCD00150329 |
Quantity | 25 g |
Synonym | lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate |
InChI Key | YECIFGHRMFEPJK-UHFFFAOYSA-N |
SMILES | O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C |
IUPAC Name | {[(2,6-dimethylphenyl)carbamoyl]methyl}diethylazanium hydrate chloride |
Molecular Weight (g/mol) | 288.82 |
PubChem CID | 16219577 |
Show More |
Safety and Handling
Product Identifier
- Lidocaine Hydrochloride
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction