Learn More
Gilac™ Lid with Handle for Round HACCP Food Contact Container
Product Code. p-200009536
White, 50 L Round HACCP Food Contact Container, Each
Click to view available options
Color:
Blue
Green
Red
White
Yellow
For Use With (Equipment):
50 L Round HACCP Food Contact Container
75 L Round HACCP Food Contact Container
Unit Size:
Each
Lid with handle for round HACCP container.
  • Lid with handle for round HACCP container.
Specifications
Product Type HACCP Food Container Lid
Material Polyethylene
Color White
For Use With (Equipment) 50 L Round HACCP Food Contact Container
Product Content Correction
Product Title
Gilac™ Lid with Handle for Round HACCP Food Contact Container >