Learn More
Eppendorf™ Li-Polymer Accumulator
Brand: Eppendorf™ 4861601000
260.39 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Specifications
Li-Polymer | |
Accumulator | |
1 Each |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now