missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX1 Monoclonal Antibody (OTI2G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504534S

This item is not returnable.
View return policy
Description
LHX1 Monoclonal antibody specifically detects LHX1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
LHX1 | |
Monoclonal | |
0.83 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P48742 | |
Mouse | |
Affinity chromatography | |
RUO | |
3975 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI2G3 | |
Unconjugated | |
Lhx1 | |
hLim-1; Homeobox protein Lim-1; LHX1; LHX-1; LIM homeo box protein 1; LIM homeobox 1; LIM homeobox protein 1; LIM/homeobox protein Lhx1; Lim1; Lim-1; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx1; MGC126723; MGC138141; rlim; transcription factor; homeobox protein | |
Human recombinant protein fragment corresponding to amino acids 100-362 of human LHX1 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
LHX1 Monoclonal Antibody (OTI2G3), TrueMAB™, OriGene
Spot an opportunity for improvement?Share a Content Correction