missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX1 Monoclonal Antibody (OTI2G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504534

This item is not returnable.
View return policy
Description
This gene encodes a member of a large protein family which contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in control of differentiation and development of neural and lymphoid cells. A similar protein in mice is an essential regulator of the vertebrate head organizer.Specifications
LHX1 | |
Monoclonal | |
0.83 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P48742 | |
LHX1 | |
Human recombit protein fragment corresponding to amino acids 100-362 of human LHX1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2G3 | |
Unconjugated | |
LHX1 | |
LIM-1, LIM1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3975 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
LHX1 Monoclonal Antibody (OTI2G3), TrueMAB™, OriGene
Spot an opportunity for improvement?Share a Content Correction