missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals [Leu8] Renin Substrate Tetradecapeptide, Rat

For cellular and molecular biology applications

577.00 NOK - 2690.00 NOK

Specifications

Color White to off-white
Form Solid
Molecular Weight (g/mol) 1789.06
Storage Requirements -30° to -10°C
View More Specs

Products 2
Product Code Brand Substrate Quantity Price Quantity & Availability  
Product Code Brand Substrate Quantity Price Quantity & Availability  
15427379
View Documents
Thermo Scientific Chemicals
J66131.#0
5mg
2690.00 NOK
5mg
Please to purchase this item. Need a web account? Register with us today!
15437379
View Documents
Thermo Scientific Chemicals
J66131.M¬
1mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

White to off-white
Solid
1789.06
-30° to -10°C
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals [Leu8] Renin Substrate Tetradecapeptide, Rat

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.