Learn More
Emma Safety Footwear Leo Safety Shoes
Includes a steel toe cap, a polyurethane outer toe cap and an easy-twist sole that provides a sticky grip. Available in a variety of widths.
1230.00 NOK
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Closure Type | Laces |
Color | Black |
Design | Leo |
Liner Material | Hydro-Tec™ Sanitized Silver |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15988821
|
Emma Safety Footwear
MM70054835 |
35 |
1230.00 NOK
1 pair |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||||
15998821
|
Emma Safety Footwear
MM70054836 |
36 |
1230.00 NOK
1 pair |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||||
15908831
|
Emma Safety Footwear
MM70054837 |
37 |
1230.00 NOK
1 pair |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||||
15918831
|
Emma Safety Footwear
MM70054838 |
38 |
1230.00 NOK
1 pair |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||||
15928831
|
Emma Safety Footwear
MM70054839 |
39 |
1230.00 NOK
1 pair |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||||
15938831
|
Emma Safety Footwear
MM70054848 |
48 |
1230.00 NOK
1 pair |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||||
15948831
|
Emma Safety Footwear
MM70054849 |
49 |
1230.00 NOK
1 pair |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||||
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather
- GripForce™ Easy Twist
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
Steel |
Laces | |
Leo | |
Leather |
Product Title
Emma Safety Footwear Leo Safety Shoes