missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Lead Reagent Test Kits
With certificate for optimum precision.
Brand: WTW™ 252034
This item is not returnable.
View return policy
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifications
For waste water, drinking water | |
Certificates | |
1 Pack | |
Photometric |
For PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex, Photoflex photometers | |
50 | |
0.010 - 5.00mg/I Pb | |
pH Test kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- lead test method: photometric 0.010 - 5.00mg/l Pb spectroquant
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H332-Harmful if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : potassium cyanide