Learn More
Buerkle™ LDPE Narrow-Necked Bottle Product Code.: 15455554
Estimated Shipment: 16-07-2025
to see stock.
Add to basket
Buerkle™ LDPE Narrow-Necked Bottle
Product Code. 15455554 Shop All Bürkle Products
20 mL
Click to view available options
Capacity (Metric):
10 mL
20 mL
30 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
Product Code. 15455554
Brand: Buerkle™ 03020020
Estimated Shipment: 16-07-2025
Buerkle™ LDPE Narrow-Necked Bottle for storing, packing, lab, taking samples, sampling and transport of liquids.
  • Universal bottles for storing, packing, lab, taking samples, sampling and transport of liquids.
  • With screw cap
Specifications
Capacity (Metric) 20 mL
Color Transparent
Diameter (Metric) Inner Neck 9 mm
For Use With (Application) Suitable For Use With Foodstuff
Material LDPE
Quantity 1
Transparency Yes
Thread Size (Metric) 14 mm
Closure Color Transparent
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 58 mm
Product Type Narrow Mouth Bottle
Closure Material LDPE
Diameter (Metric) 32 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Product Title
Buerkle™ LDPE Narrow-Necked Bottle >