Learn More
Kartell™ Latex Gas Sampling Balloon
Gass collection bags that are constructed from latex. Kartell™ Latex Gas Sampling Balloon are best suited for collecting, containing, and carrying gas samples.
Brand:  Kartell™ 0123400
351.59 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 10263183
  • 418.00 NOK / Each
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Description
  • Cost effective
  • Ideal for collecting, containing, and carrying gas samples
Specifications
Specifications
Gas Sampling Balloon
5 L
Latex
230 mm
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
Product Content Correction
Product Title
Kartell™ Latex Gas Sampling Balloon > Capacity, Metric: 5L