Learn More
Kartell™ Latex Dropping Bulb with Rim
Kartell™ Latex Dropping Bulb with Rim allows for easy dispensing of liquids and can be used with Ranvier Dropping Tubes and Pasteur Pipettes.
Brand: Kartell™ 0391400
657.81 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
This item is not returnable.
View return policy
Description
- Suitable for Ranvier dropping tubes
- Suitable for Pasteur pipettes
- Constructed of latex
Specifications
Latex | |
For Ranvier Dropping Tubes and Pasteur Pipettes | |
40 mm |
Dropping Bulb | |
With Rim |
Product Content Correction
Product Title
Kartell™ Latex Dropping Bulb with Rim > Material: Latex