Learn More
Brady™ Laminated Polyester ADR 1.5 1 Explosive Substances Transport Signs
Brand: Brady™ 223595
This item is not returnable.
View return policy
Specifications
ADR 1.5RL-100*100-B7541 | |
Laminated Polyester | |
Permanent Acrylic | |
0.099 mm | |
-40 °C-110 °C | |
1.5 1 |
Gloss | |
250/Pk. | |
100 mm | |
100.00 mm | |
Diamond | |
Black on Orange |
Product Content Correction
Product Title
Brady™ Laminated Polyester ADR 1.5 1 Explosive Substances Transport Signs > Dimensions (W x H): 100 x 100mm, 250Pack