Learn More
Liebherr Laboratory performance freezer including drawers
Brand: Liebherr SFFFG4001740
39535.45 NOK valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Laboratory performance freezer including drawers
This item is not returnable.
View return policy
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%
Product Content Correction
Product Title
Liebherr Laboratory performance freezer including drawers >